Membrane filters, type 113
Supplier: Sartorius
About this item
Cellulose nitrate hydrophilic membrane filters. For sample pretreatment, particle testing and chemotaxis.
* Sterile
56 Options Available Below
Product Details & Documents
Cellulose nitrate hydrophilic membrane filters. For sample pretreatment, particle testing and chemotaxis.
* Sterile
Specifications for Product Family
Specifications
- Catalog No:
- SART11327-25-N
- 512-5116
- 512-5216
- 512-5843
- 512-5316
- 611-0675
- 512-5416
- 512-5876
- 611-0677
- 512-5917
- 514-0754
- 512-5516
- SART11306-100-N
- SART11305-13-N
- 512-5215
- 512-5315
- SART11305-47-ACN
- 512-5415
- 512-5875
- SART11305-100-N
- 512-5615
- 512-5214
- 512-5842
- 512-5314
- 512-5864
- 512-5414
- 512-5874
- 512-5915
- SART11304-100-G
- 512-5213
- 512-5313
- 512-5863
- 512-5413
- 512-5914
- 512-5513
- 512-5112
- 512-5212
- 512-5312
- 512-5412
- SART11302-90-G
- 512-5111
- 512-5211
- 512-5311
- 512-5411
- SART11342-90-G
- 512-5110
- 512-5210
- 514-0752
- 512-5861
- 512-5310
- 512-5862
- 512-5410
- 512-5871
- SART11301-70-G
- 512-5510
- SART11301-100-N
- SART11327-47-N
- Type:
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- 113
- Pore size:
- 0,20 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,65 µm
- 0,65 µm
- 0,65 µm
- 0,65 µm
- 0,65 µm
- 0,65 µm
- 0,65 µm
- 0,65 µm
- 0,80 µm
- 0,80 µm
- 0,80 µm
- 0,80 µm
- 0,80 µm
- 0,80 µm
- 0,80 µm
- 0,80 µm
- 1,20 µm
- 1,20 µm
- 1,20 µm
- 1,20 µm
- 1,20 µm
- 1,20 µm
- 3,00 µm
- 3,00 µm
- 3,00 µm
- 3,00 µm
- 3,00 µm
- 5,00 µm
- 5,00 µm
- 5,00 µm
- 5,00 µm
- 5,00 µm
- 8,00 µm
- 8,00 µm
- 8,00 µm
- 8,00 µm
- 8,00 µm
- 3,00 µm
- 8,00 µm
- 8,00 µm
- 8,00 µm
- 8,00 µm
- 8,00 µm
- 0,20 µm
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Thickness:
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- 130 µm
- Water flow rate:
- 25 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 70 ml/min/cm² (1 bar)
- 130 ml/min/cm² (1 bar)
- 130 ml/min/cm² (1 bar)
- 130 ml/min/cm² (1 bar)
- 130 ml/min/cm² (1 bar)
- 130 ml/min/cm² (1 bar)
- 130 ml/min/cm² (1 bar)
- 130 ml/min/cm² (1 bar)
- 130 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 200 ml/min/cm² (1 bar)
- 430 ml/min/cm² (1 bar)
- 430 ml/min/cm² (1 bar)
- 430 ml/min/cm² (1 bar)
- 430 ml/min/cm² (1 bar)
- 430 ml/min/cm² (1 bar)
- 570 ml/min/cm² (1 bar)
- 570 ml/min/cm² (1 bar)
- 570 ml/min/cm² (1 bar)
- 570 ml/min/cm² (1 bar)
- 570 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 430 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 750 ml/min/cm² (1 bar)
- 25 ml/min/cm² (1 bar)
- Airflow rate:
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- Max. temperature:
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- Environmental certification:
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
Specifications
Product Family Options
Product Information
- TypePore sizeØSterileAvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.SART11327-25-NType113Pore size0,20 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate25 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5116Type113Pore size0,45 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5216Type113Pore size0,45 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5843Type113Pore size0,45 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5316Type113Pore size0,45 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.611-0675Type113Pore size0,45 µmEnvironmentally PreferableThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5416Type113Pore size0,45 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5876Type113Pore size0,45 µmEnvironmentally PreferableThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.611-0677Type113Pore size0,45 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5917Type113Pore size0,45 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.514-0754Type113Pore size0,45 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5516Type113Pore size0,45 µmEnvironmentally PreferableThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
More
Specifications
Cat. No.SART11306-100-NType113Pore size0,45 µmEnvironmentally PreferableThickness130 µmWater flow rate70 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.SART11305-13-NType113Pore size0,65 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate130 ml/min/cm² (1 bar)Airflow rateMax. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5215Type113Pore size0,65 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate130 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5315Type113Pore size0,65 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate130 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.SART11305-47-ACNType113Pore size0,65 µmEnvironmentally PreferableThickness130 µmWater flow rate130 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5415Type113Pore size0,65 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate130 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5875Type113Pore size0,65 µmEnvironmentally PreferableThickness130 µmWater flow rate130 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.SART11305-100-NType113Pore size0,65 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate130 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - 1130,65 µm142 mm-Discontinued-Specifications:
More
Specifications
Cat. No.512-5615Type113Pore size0,65 µmEnvironmentally PreferableThickness130 µmWater flow rate130 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5214Type113Pore size0,80 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5842Type113Pore size0,80 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5314Type113Pore size0,80 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5864Type113Pore size0,80 µmEnvironmentally PreferableThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5414Type113Pore size0,80 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5874Type113Pore size0,80 µmEnvironmentally PreferableThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
More
Specifications
Cat. No.512-5915Type113Pore size0,80 µmEnvironmentally PreferableThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.SART11304-100-GType113Pore size0,80 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5213Type113Pore size1,20 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5313Type113Pore size1,20 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5863Type113Pore size1,20 µmEnvironmentally PreferableThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5413Type113Pore size1,20 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5914Type113Pore size1,20 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5513Type113Pore size1,20 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate200 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5112Type113Pore size3,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate430 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5212Type113Pore size3,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate430 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5312Type113Pore size3,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate430 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5412Type113Pore size3,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate430 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.SART11302-90-GType113Pore size3,00 µmEnvironmentally PreferableThickness130 µmWater flow rate430 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5111Type113Pore size5,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate570 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5211Type113Pore size5,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate570 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5311Type113Pore size5,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate570 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5411Type113Pore size5,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate570 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.SART11342-90-GType113Pore size5,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate570 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5110Type113Pore size8,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5210Type113Pore size8,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.514-0752Type113Pore size8,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5861Type113Pore size8,00 µmEnvironmentally PreferableThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5310Type113Pore size8,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5862Type113Pore size3,00 µmEnvironmentally PreferableThickness130 µmWater flow rate430 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5410Type113Pore size8,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.512-5871Type113Pore size8,00 µmEnvironmentally PreferableThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.SART11301-70-GType113Pore size8,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.512-5510Type113Pore size8,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.SART11301-100-NType113Pore size8,00 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate750 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.SART11327-47-NType113Pore size0,20 µmEnvironmentally PreferableLow Manufacturing ImpactThickness130 µmWater flow rate25 ml/min/cm² (1 bar)Airflow rate-Max. temperature130 °CEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact