Order Entry
Belgium
ContactUsLinkComponent
Hand cleansing foam, Solopol® GrittyFOAM
Hand cleansing foam, Solopol® GrittyFOAM
Catalog # 115-2087
Supplier:  Deb-STOKO
undefined
Hand cleansing foam, Solopol® GrittyFOAM
Catalog # 115-2087
Supplier:  Deb-STOKO

Specifications

  • Description:
    Hand cleansing foam, Solopol® GrittyFOAM, 3250 ml cartridge
  • Pk:
    4

Specifications

About this item

Medium heavy foam hand cleanser with scrubbing agent to remove medium to heavy contamination, e.g. oils, greases, carbon black, lubricants. For use in industrial environments.

  • Contains non abrasive scrubbing agent
  • 3 ml of Solopol GrittyFOAM liquid transforms into 15 ml of foam, providing up to 47% more hand washes per litre compared to traditional hand cleansers
  • Formulation leaves hands feeling smooth and conditioned
  • Petroleum solvent-free, silicone-free, colourant-free
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904