Orders LinkContactUsLinkComponent
Test tubes
Supplier:   DWK Life Sciences
Test tubes
Supplier:  DWK Life Sciences
Certificates
8 Options Available Below
Product Details & Documents
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide variety of applications.
Product Family Options
Without rim
Product Information
  • Length
    Ext.Ø
    Wall thickness
    Availability
    Price
  • 100 mm
    12 mm
    1,1 - 1,2 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 49,70 €
    49,70 €
    1 * 100 ST
       49,70 €
  • 100 mm
    16 mm
    1,1 - 1,2 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 81,20 €
    81,20 €
    1 * 100 ST
       81,20 €
  • 160 mm
    16 mm
    1,1 - 1,2 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 76,60 €
    76,60 €
    1 * 100 ST
       76,60 €
  • 180 mm
    18 mm
    1,1 - 1,2 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 114,00 €
    114,00 €
    1 * 100 ST
       114,00 €
  • 200 mm
    30 mm
    1,1 - 1,2 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 76,10 €
    76,10 €
    1 * 50 ST
       76,10 €
With rim
Product Information
  • Length
    Ext.Ø
    Wall thickness
    Availability
    Price
  • 100 mm
    12 mm
    0,9 - 1,0 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 42,30 €
    42,30 €
    1 * 100 ST
       42,30 €
  • 160 mm
    16 mm
    1,1 - 1,2 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 76,60 €
    76,60 €
    1 * 100 ST
       76,60 €
  • 200 mm
    25 mm
    1,1 - 1,2 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 2 660,00 €
    2 660,00 €
    1 * 1.000 ST
       2 660,00 €