Automated staining coverslipper, Combistainer
Supplier: SERMO INDUSTRIALISATION
Certificates
About this item
Combistainer, a fully automated stainer and taper for up to ten protocols simultaneously: Staining, dewaxing and dehydration.
- Tactile screen, password security
- Integrated reagents management software
- Barcode reader for slide process management (USB connection) (optional)
- Manual slide rack entrance for cover slipping of IHC slides
- Adjustable xylene dispensing volume
- Automated transfer to drying unit: Forced air drying chamber up to seven slide racks
Combistainer provides automation features for histology laboratories in both staining and coverslipping applications with KP Tape film.
Capacity of Automated staining coverslipper:
Stainer: Three positions - Two slide racks of 20 slides
- Two drying stations of four racks
- Two water baths of four racks
- 27 staining troughs for two racks
Size of Automated staining coverslipper:
1178×769×660 mm (stainer) ;762×519×660 mm (taper)
Frequently Bought Together