Order Entry
Belgium
Orders LinkContactUsLinkComponent
Magnesium standard solution

Magnesium standard solution

Magnesium standard solution, 100 ml (NIST), 1000 mg/l as Mg, 100 mL
Catalog # HACH1479442
Supplier:   Hach
Magnesium standard solution
Catalog # HACH1479442
Supplier:  Hach
CAS Number:  
Magnesium standard solution
Magnesium standard solution

Magnesium standard solution, 100 ml (NIST), 1000 mg/l as Mg, 100 mL

CAS Number:  
Price
50,50 €
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solutions for accuracy checks in magnesium determinations. HNO₃ matrix.

Specifications

Specifications

  • Concentration:
    1000 mg/l as Mg
  • Pk:
    100 mL
  • Description:
    Magnesium standard solution, 100 ml (NIST)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.