Quaternary ammonium compound (QAC) standard solution
Quaternary ammonium compound (QAC) standard solution, 100 ml, 100 mg/l as CTAB, 100 mL
Catalog # HACH2415342
Supplier: Hach
Quaternary ammonium compound (QAC) standard solution
Catalog # HACH2415342
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
For accuracy checks in quaternary ammonium compound (QAC) determinations.
Specifications
Specifications
- Concentration:100 mg/l as CTAB
- Pk:100 mL
- Description:Quaternary ammonium compound (QAC) standard solution, 100 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



