Order Entry
Belgium
Orders LinkContactUsLinkComponent
Barium standard solution

Barium standard solution

Barium, Ba, 1000 mg/L, H₂O, 100 mL
Catalog # HACH1461142
Supplier:   Hach
Barium standard solution
Catalog # HACH1461142
Supplier:  Hach
CAS Number:  
Barium standard solution
Barium standard solution

Barium, Ba, 1000 mg/L, H₂O, 100 mL

CAS Number:  
Price
38,80 €
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in barium determinations.

Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Element:
    Barium
  • Symbol:
    Ba

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.