Order Entry
Belgium
Orders LinkContactUsLinkComponent
Standard solution, detergent

Standard solution, detergent

Standard solution, detergent, pack of 16×10 ml voluette ampoules, 60 mg/l as LAS, 16
Catalog # 705-2016
Supplier:   Hach
Standard solution, detergent
Catalog # 705-2016
Supplier:  Hach
CAS Number:  
Standard solution, detergent
Standard solution, detergent

Standard solution, detergent, pack of 16×10 ml voluette ampoules, 60 mg/l as LAS, 16

CAS Number:  
Price
70,10 €
Log in for availability
1 * 16 ST
Product Details & Documents

For accuracy checks in detergents (surfactants) determinations.

Specifications

Specifications

  • Concentration:
    60 mg/l as LAS
  • Pk:
    16
  • Description:
    Standard solution, detergent, pack of 16×10 ml voluette ampoules

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.