Order Entry
Belgium
Orders LinkContactUsLinkComponent
Silver nitrate standard solution

Silver nitrate standard solution

Silver nitrate standard solution, 0,141 N, 500 ml, 500 mL
Catalog # HACH1255149
Supplier:   Hach
Silver nitrate standard solution
Catalog # HACH1255149
Supplier:  Hach
CAS Number:  
Silver nitrate standard solution
Silver nitrate standard solution

Silver nitrate standard solution, 0,141 N, 500 ml, 500 mL

CAS Number:  
Price
79,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

Standard solution for chloride determination by argentometric burette titration.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Silver nitrate standard solution, 0,141 N, 500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.