Media in bottles, wort agar
DOEH840360782
:
- Pack type:Bottle
- Pk:9
- Description:Wort agar, glas bottle
- Size:250 ml
- Filling:250 ml
Agar for isolation yeast and moulds.
- For membrane filtration, pour plate and streaking method
- High selectivity