Order Entry
Belgium
ContactUsLinkComponent
Iodine 0.05 mol/l (0.1N), BAKER ANALYZED® volumetric solution, J.T.Baker®
Irritant
Health
Iodine 0.05 mol/l (0.1N), BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5623.1000
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Health
Iodine 0.05 mol/l (0.1N), BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5623.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.0995 - 0.1005
  • Normality:
    0.0995 - 0.1005

Specification Test Results

About this item

 :
 :253.81 g/mol
ADR:8,III
 :GHS07
 :GHS08
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355