Order Entry
Belgium
ContactUsLinkComponent
Silica gel, BAKER ANALYZED® (4-10 Mesh) desiccant, with moisture indicator, J.T.Baker®
Toxic
Health
Environment
Silica gel, BAKER ANALYZED® (4-10 Mesh) desiccant, with moisture indicator, J.T.Baker®
Catalog # 3402-01
Supplier:  Avantor
CAS Number:  
undefined
Toxic
Health
Environment
Silica gel, BAKER ANALYZED® (4-10 Mesh) desiccant, with moisture indicator, J.T.Baker®
Catalog # 3402-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth, black T/E cap
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≤ 6.0 %
  • Loss on Heating at 200°C:
    ≤ 6.0 %
  • Suitability for Moisture Absorption at 80% Relative Humidity:
    ≥ 33 %
  • Suitability for Moisture Absorption at 20% Relative Humidity:
    ≥ 10 %
  • Suitability as Indicating Desiccant:
    Passes Test
  • Mesh - Thru U.S. No. 10 Sieve:
    ≤ 5.0 %

Specification Test Results

About this item

 :
 :60.08 g/mol
Density:2.10 kg/m³
 :GHS06
 :GHS08
 :GHS09
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232