Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Polysorbate 80, J.T.Baker®

Supplier:   Avantor
Polysorbate 80, J.T.Baker®
Supplier:  Avantor
Certificates
Polysorbate 80, J.T.Baker®

2 Options Available Below

Product Details & Documents

Polyoxyethylene (20) Sorbitan Monooleate

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Storage Temperature:Ambient
  • EINECS:500-018-3

 

Specifications for Product Family

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Identification (by IR):
    Passes Test
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle, amber, narrow mouth
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L

  • Jerrican, poly pail
    19 L
    Log in for availability
    Log in for availability
    1 * 19 L
    1 * 19 L

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.