Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
Carbon disulphide ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
 
 
 
Carbon disulphide ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Meets ACS Reagent Chemical Requirements:
    = 99.9 %
  • For Laboratory,Research,or Manufacturing Use:
    = 10
  • Assay (CS₂) (by GC, corrected for water):
    ≥99.9 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.001 %
  • Hydrogen Sulfide (H₂S):
    Passes Test
  • Sulfur Dioxide (SO₂):
    Passes Test
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %

 

 

Frequently Bought Together