Magnesium sulphate, anhydrous ≥99%, powder, BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7487-88-9
:
Formula:MgSO₄
MDL Number:MFCD00011110
:120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥99.0%
- Assay (MgSO₄) (ignited basis):≥99.0 %
- Loss on Ignition:≤2.0 %
- Chloride (Cl):≤0.001 %
- Nitrate (NO₃):≤0.004 %
- Ammonium (NH₄):≤0.004 %
- Calcium (Ca):≤0.06 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Manganese (Mn):≤0.001 %
- Trace Impurities - Arsenic (As):≤1 ppm
Specification Test Results
Frequently Bought Together