Order Entry
Belgium
ContactUsLinkComponent
 
Magnesium chloride hexahydrate 98.0-101.0% USP, Macron Fine Chemicals™
 
undefined
Magnesium chloride hexahydrate 98.0-101.0% USP, Macron Fine Chemicals™
 

 

  • Meets U.S.P Requirements:
  • GMP Manufactured Product:
  • Preserve in Tight Containers:
  • CAUTION: For Manufacturing, processing or repackaging:
    98.0 - 101.0 %
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (MgCl₂·6H₂O):
    98.0 - 101.0 %
  • USP - Barium (Ba):
    Passes Test
  • USP - Calcium (Ca):
    ≤0.01 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Insoluble Matter:
    ≤0.005 %
  • USP - pH of Aqueous Solution:
    4.5 - 7.0
  • USP - Potassium (K):
    Passes Test
  • USP - Sulfate (SO₄):
    ≤0.005 %

 

 

Frequently Bought Together