Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
 
n-Heptane ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™
 
undefined
 
 
 
 
n-Heptane ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™
 

 

  • For Laboratory,Research,or Manufacturing Use:
    ≥99.0%
  • Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water):
    ≥99.0 %
  • Acidity (as CH₃COOH):
    Passes Test
  • Residue after Evaporation:
    ≤0.001 %
  • Sulfur Compounds (as S):
    ≤0.005 %
  • Specific Gravity at 60°/60°F:
    0.68 - 0.72
  • Thiophene:
    Passes Test

 

 

Frequently Bought Together