1-Butanol ≥99.4%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9054-R
Supplier: Avantor
CAS Number:
Specifications
- Pk:370 lb
- Seq:7266
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water):≥99.4 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.0050 %
- Titrable Acid (meq/g):≤0.0008
- Butyl Ether (C₈H₁₈O)(by GC):≤0.2 %
- Carbonyl Compounds - Butyraldehyde:≤0.01 %
- Water (by KF, coulometric):≤0.1 %
- Meets Reagent Specifications for testing USP/NF monographs:—
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:13,01539
Boiling Pt:117.6 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:37 °C
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
:GHS02
:GHS08
:GHS07
CAS Number:71-36-3
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964