Order Entry
Belgium
ContactUsLinkComponent
Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9324-R
Supplier:  Avantor
CAS Number:  
undefined
Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9324-R
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    600 lb
  • Seq:
    7350
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (CH₂Cl₂) (by GC, corrected for water):
    ≥ 99.5 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 0.0020 %
  • Titrable Acid (µeq/g):
    ≤ 0.3
  • Free Halogens:
    Passes Test
  • Water (by KF, coulometric):
    ≤ 0.02 %
  • Appearance (Clear, colorless liquid):
    39.5 °C
  • Boiling Point (typical)................. 39.5 °C.:
    1.318

Specification Test Results

About this item

Synonyms: Methylene chloride
 :
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :84.93 g/mol
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881