Order Entry
Belgium
ContactUsLinkComponent
 
Reverse osmosis water purification system, PURELAB® Chorus 3
 
undefined
Reverse osmosis water purification system, PURELAB® Chorus 3

 

  • Catalog No:
  • ELGARO330BPM1
  • ELGARO320XXM1
  • ELGARO330XXM1
  • ELGARO320BPM1
  • ELGARO310XXM1
  • ELGARO310BPM1
  • Model:
  • PURELAB® Chorus 3
  • PURELAB® Chorus 3
  • PURELAB® Chorus 3
  • PURELAB® Chorus 3
  • PURELAB® Chorus 3
  • PURELAB® Chorus 3
  • Average daily needs:
  • 720 l
  • 480 l
  • 720 l
  • 480 l
  • 240 l
  • 240 l
  • Feed water:
  • Potable mains water supply
  • Potable mains water supply
  • Potable mains water supply
  • Potable mains water supply
  • Potable mains water supply
  • Potable mains water supply
  • Production flow rate:
  • 30 L/h
  • 20 L/h
  • 30 L/h
  • 20 L/h
  • 10 L/h
  • 10 L/h
  • TOC content:
  • <50 ppb
  • <50 ppb
  • <50 ppb
  • <50 ppb
  • <50 ppb
  • <50 ppb
  • Inorganics @ 25°C:
  • > 95% rejection
  • > 95% rejection
  • > 95% rejection
  • > 95% rejection
  • > 95% rejection
  • > 95% rejection
  • Bacteria:
  • <50 CFU/ml
  • <50 CFU/ml
  • <50 CFU/ml
  • <50 CFU/ml
  • <50 CFU/ml
  • <50 CFU/ml
  • W×D×H:
  • 376×340×435 mm
  • 376×340×435 mm
  • 376×340×435 mm
  • 376×340×435 mm
  • 376×340×435 mm
  • 376×340×435 mm
  • Weight:
  • 17 kg (19 kg with boost pump)
  • 16 kg (18 kg with boost pump)
  • 17 kg (19 kg with boost pump)
  • 16 kg (18 kg with boost pump)
  • 15 kg (17 kg with boost pump)
  • 15 kg (17 kg with boost pump)

 

Frequently Bought Together