ContactUsLinkComponent
Micro spatula for NMR sample tubes, SP Wilmad-LabGlass
Micro spatula for NMR sample tubes, SP Wilmad-LabGlass
Catalog # WILM806
Supplier:  SP Industries
Micro spatula for NMR sample tubes, SP Wilmad-LabGlass
Catalog # WILM806
Supplier:  SP Industries
Specifications
  • Length:
    250 mm
  • Blade width:
    3 mm
  • Pk:
    1
Specifications
About this item
Stainless steel. This spatula fits into the most common NMR tubes. It enables users to get a tiny powder sample into any tube larger than 4 mm ext. Ø.
Ideal for adding or removing samples (including freeze-dried). Scoop samples with the round end or pry stiff samples with the sharp-edged flat end.