ContactUsLinkComponent
Test tube racks, SP Wilmad-LabGlass
Test tube racks, SP Wilmad-LabGlass
Catalog # WILM820-A
Supplier:  SP Industries
Test tube racks, SP Wilmad-LabGlass
Catalog # WILM820-A
Supplier:  SP Industries
Specifications
  • For tube Ø:
    5 mm
  • No. of Holes:
    72
  • Pk:
    1
Specifications
About this item
PP, natural.
These lightweight racks maximise benchtop storage capacity and is virtually inert. Ideal for use with NMR and EPR tubes.