Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Conductivity standard, 12,88 mS/cm, 250 mL
Catalog # METT51350094
Supplier:   Mettler - Toledo
Conductivity standards
Catalog # METT51350094
Supplier:  Mettler - Toledo
CAS Number:  
Conductivity standards
Conductivity standards
Conductivity standard, 12,88 mS/cm, 250 mL
CAS Number:  
Price
31,60 €
Log in for availability
1 * 250 mL
Product Details & Documents
A slight change in temperature usually has a big impact on the conductivity value of a standard solution. A table on every bottle label indicates the conductivity values at the most common measurement temperatures.
During calibration the meter automatically references to this table for temperature compensation. If possible, calibration and measurements should be performed at the same temperature
Documents
Certificates
Search by Lot Number
Specifications
Specifications
  • Pk:
    250 mL
  • Description:
    Conductivity standard
  • Conductivity:
    12,88 mS/cm