ContactUsLinkComponent
Turbidity standards for Turbicheck
Turbidity standards for Turbicheck
Catalog # 705-0934
Supplier:  Lovibond
CAS Number:  
Turbidity standards for Turbicheck
Catalog # 705-0934
Supplier:  Lovibond
CAS Number:  
Specifications
  • Pk:
    1 SET
  • Turbidity:
    200 NTU
    800 NTU
    <0,1 NTU
    20 NTU
  • Description:
    T-Cal, set of 4 turbidity standards
  • Pk Info:
    4 sealed cells