Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffer solutions

pH buffer solutions
 
pH buffer solutions
pH buffer solutions

 

Product Family Options

 

  • Description
    pH value
    Colour
    Pk Info
    Pk
     
     
  • Technical buffer solution, 30×20 ml
    2,00
    30
       
       
    1 * 30 ST
    1 * 30 ST - 79,80 €
    79,80 €
    1 * 30 ST
       79,80 €

  • Technical buffer solution, 250 ml
    2,00
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €

  • Technical buffer solution, 250 ml
    4,01
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €

  • Technical buffer solution, 6×250 ml
    4,01
    6
       
       
    1 * 6 ST
    1 * 6 ST - 91,10 €
    91,10 €
    1 * 6 ST
       91,10 €

  • Technical buffer solution, 250 ml
    7,00
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €

  • Technical buffer solution, 250 ml
    9,21
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €

  • Technical buffer solution, 6×250 ml
    9,21
    Colourless
    250 ml
    6
       
       
    1 * 6 ST
    1 * 6 ST - 91,10 €
    91,10 €
    1 * 6 ST
       91,10 €

  • Technical buffer solution, 250 ml
    10,00
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €

  • Technical buffer solution, 6×250 ml
    10,00
    6
       
       
    1 * 6 ST
    1 * 6 ST - 91,10 €
    91,10 €
    1 * 6 ST
       91,10 €

  • Technical buffer solution, 30×20 ml
    11,00
    30
       
       
    1 * 30 ST
    1 * 30 ST - 79,80 €
    79,80 €
    1 * 30 ST
       79,80 €

  • Technical buffer solution, 250 ml
    11,00
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €

  • Technical buffer solution, 6×250 ml
    4,64
    Colourless
    250 ml
    6