Magnetic stirring bars, polygon, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
PTFE covered with strong Alnico V magnetic core. The multifaceted surfaces add turbulence relative to smooth cylindrical bars of a similar size.
- Autoclave safe
- FDA grade PTFE is durable and inert
- Pivot ring minimises the contact area between the bar and the vessel
Frequently Bought Together