Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer tablets, Lovibond®

Buffer tablets, Lovibond®

Supplier:   Lovibond
Buffer tablets, Lovibond®
Supplier:  Lovibond
Certificates
Buffer tablets, Lovibond®

6 Options Available Below

Product Details & Documents

Ideal for preparing buffer solutions of 4, 7 and 10 pH units.

pH tablet can be added to 100 ml of distilled or deionised water for preparing solution of specified concentration.

Product Family Options

Product Information

  • pH value
    Pk
    Availability
    Price
  • 4
    100 Tab.
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 27,70 €
    27,70 €
    1 * 1 EA
       27,70 €

  • 4
    250 Tab.
    Log in for availability
    Log in for availability
    1 * 250 Tablet
    1 * 250 Tablet - 56,80 €
    56,80 €
    1 * 250 Tablet
       56,80 €

  • 7
    100 Tab.
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 27,70 €
    27,70 €
    1 * 1 EA
       27,70 €

  • 7
    250 Tab.
    Log in for availability
    Log in for availability
    1 * 250 Tablet
    1 * 250 Tablet - 56,80 €
    56,80 €
    1 * 250 Tablet
       56,80 €

  • 10
    100 Tab.
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 27,70 €
    27,70 €
    1 * 1 EA
       27,70 €

  • 10
    250 Tab.
    Log in for availability
    Log in for availability
    1 * 250 Tablet
    1 * 250 Tablet - 56,80 €
    56,80 €
    1 * 250 Tablet
       56,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.