Order Entry
Belgium
ContactUsLinkComponent
proBDNF Rapid™ ELISA Kit: Human, Mouse, Rat
proBDNF Rapid™ ELISA Kit: Human, Mouse, Rat
Supplier:  Biosensis
Certificates
undefined
proBDNF Rapid™ ELISA Kit: Human, Mouse, Rat
Supplier:  Biosensis

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
    Rat
    Mouse
  • Target protein:
    proBDNF
  • Cross reactivity:
    Due to a high degree of sequence homology, the human proBDNF ELISA kit cross-reacts with the mouse and rat form of proBDNF. Other species have not yet been tested, but cross-reactivity with a wide range of mammalian forms of proBDNF is expected. A cross-reactivity of 2% has been observed for mature BDNF (assayed at 25 ng/ml). The antibodies do not cross-react with nerve growth factor (NGF), neurotrophin-3 (NT-3) or NT-4/5.
  • Detection range:
    15,6 – 1000 pg/ml
  • Sensitivity:
    Typical limit of detection (LOD) for proBDNF is 30 pg/ml determined as 150% of the blank value

Specifications

Frequently Bought Together