Order Entry
Belgium
Orders LinkContactUsLinkComponent
NGF Rapid™ ELISA kit: human

NGF Rapid™ ELISA kit: human

Supplier:   Biosensis
NGF Rapid™ ELISA kit: human
Supplier:  Biosensis
Certificates
NGF Rapid™ ELISA kit: human

2 Options Available Below

Product Details & Documents

The Biosensis NGF Rapid™ enzyme-linked immunosorbent assay (ELISA) kit is a sandwich ELISA that allows the quantification of human NGF in less than 3 hours in cell culture supernatants, serum, plasma (citrate) and brain extracts only if used as directed.

This NGF Rapid™ ELISA Kit is designed to measure human NGF and thus employs a recombinant human NGF standard approved by the World Health Organization (WHO). Due to a high degree of NGF sequence homology, the antibodies used in this kit will also detect NGF from other species including mouse and rat.

The purpose of this kit is the in vitro quantitative determination of NGF in samples such as serum, plasma, cell culture supernatants and brain extracts.

The antibodies used in this ELISA kit bind epitopes within the mature domain of the protein and therefore can recognize both the pro- and the mature form of NGF. However, internal validation data demonstrates <0,1% cross-reactivity (determined at 25 ng/ml, diluted in assay buffer) with human proNGF protein, suggesting the preferential quantification of mature NGF over full-length human proNGF. Note that accurate NGF quantification in human citrate-plasma requires the addition of Heterophilic Antibody Blocker provided in the kit, and available for purchase separately.

This kit consists of a pre-coated mouse monoclonal anti-NGF capture antibody, a biotinylated anti-NGF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a coloured reaction product which is directly proportional to the concentration of NGF present in samples and protein standards. A human NGF positive control (QC sample) is provided to assure consistent assay performance.

Please refer to the kit protocol for specific use instructions for each substrate application, in particular serum and plasma samples.

Delivery information: Each kit contains 1 or 2×96 pre-coated strip plates, protein standards, detection reagents, wash and sample buffers and detailed instructions.

Caution: For research use only. Not for diagnostic or clinical purposes.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • BSENBEK-2212-1P
  • BSENBEK-2212-2P
  • Target protein:
  • NGF
  • NGF
  • Primary antibody reactivity:
  • Human
  • Human
  • Assay Type:
  • Sandwich
  • Sandwich
  • Format:
  • Pre-coated
  • Pre-coated
  • Sensitivity:
  • Typical limit of detection (LOD) for human NGF is 2 pg/ml determined as 150% of the blank value
  • Typical limit of detection (LOD) for human NGF is 2 pg/ml determined as 150% of the blank value
  • Detection range:
  • 3,9 – 250 pg/ml
  • 3,9 – 250 pg/ml
  • Cross reactivity:
  • The antibodies used in this ELISA kit bind epitopes within the mature domain of the protein and therefore can recognise both the pro- and the mature form of NGF. However, human proNGF protein shows <0,1% cross-reactivity (determined at 25 ng/ml) allowing specific quantification of mature NGF. This NGF ELISA does not cross-react with brain derived neurotrophic factor (BDNF), neurotrophin-3 (NT-3), NT-4/5, glial cell line-derived neurotrophic factor (GDNF) and vascular endothelial growth factor (VEGF165) tested at 25 ng/ml. Due to a high degree of NGF sequence homology, the antibodies used in this kit will also detect NGF from other species including mouse and rat.
  • The antibodies used in this ELISA kit bind epitopes within the mature domain of the protein and therefore can recognise both the pro- and the mature form of NGF. However, human proNGF protein shows <0,1% cross-reactivity (determined at 25 ng/ml) allowing specific quantification of mature NGF. This NGF ELISA does not cross-react with brain derived neurotrophic factor (BDNF), neurotrophin-3 (NT-3), NT-4/5, glial cell line-derived neurotrophic factor (GDNF) and vascular endothelial growth factor (VEGF165) tested at 25 ng/ml. Due to a high degree of NGF sequence homology, the antibodies used in this kit will also detect NGF from other species including mouse and rat.

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • BSENBEK-2212-1P
    NGF Rapid™ ELISA Kit: Human, 1×96-well plate
    NGF Rapid™ ELISA Kit: Human, 1×96-well plate
    96 Tests
    Log in for availability
    Log in for availability
    1 * 96 Tests
    1 * 96 Tests - 424,00 €
    424,00 €
    1 * 96 Tests
       424,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BSENBEK-2212-1P
    Target protein
    NGF
    Primary antibody reactivity
    Human
    Assay Type
    Sandwich
    Format
    Pre-coated
    Sensitivity
    Typical limit of detection (LOD) for human NGF is 2 pg/ml determined as 150% of the blank value
    Detection range
    3,9 – 250 pg/ml
    Cross reactivity
    The antibodies used in this ELISA kit bind epitopes within the mature domain of the protein and therefore can recognise both the pro- and the mature form of NGF. However, human proNGF protein shows <0,1% cross-reactivity (determined at 25 ng/ml) allowing specific quantification of mature NGF. This NGF ELISA does not cross-react with brain derived neurotrophic factor (BDNF), neurotrophin-3 (NT-3), NT-4/5, glial cell line-derived neurotrophic factor (GDNF) and vascular endothelial growth factor (VEGF165) tested at 25 ng/ml. Due to a high degree of NGF sequence homology, the antibodies used in this kit will also detect NGF from other species including mouse and rat.
  • BSENBEK-2212-2P
    NGF Rapid™ ELISA Kit: Human, 2×96-well plate
    NGF Rapid™ ELISA Kit: Human, 2×96-well plate
    192 Tests
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 772,00 €
    772,00 €
    1 * 1 EA
       772,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BSENBEK-2212-2P
    Target protein
    NGF
    Primary antibody reactivity
    Human
    Assay Type
    Sandwich
    Format
    Pre-coated
    Sensitivity
    Typical limit of detection (LOD) for human NGF is 2 pg/ml determined as 150% of the blank value
    Detection range
    3,9 – 250 pg/ml
    Cross reactivity
    The antibodies used in this ELISA kit bind epitopes within the mature domain of the protein and therefore can recognise both the pro- and the mature form of NGF. However, human proNGF protein shows <0,1% cross-reactivity (determined at 25 ng/ml) allowing specific quantification of mature NGF. This NGF ELISA does not cross-react with brain derived neurotrophic factor (BDNF), neurotrophin-3 (NT-3), NT-4/5, glial cell line-derived neurotrophic factor (GDNF) and vascular endothelial growth factor (VEGF165) tested at 25 ng/ml. Due to a high degree of NGF sequence homology, the antibodies used in this kit will also detect NGF from other species including mouse and rat.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.