Orders LinkContactUsLinkComponent
Adapters, sperical joints
Adapters, sperical joints
Adapters, sperical joints
Adapters, sperical joints
For reaction vessels, 316 stainless steel, thread length 14 mm.
Product Family Options
  • Type
  • Socket S 35 with steel thread M 33×1,5, O.Ø 30×2,4 mm, I.Ø 25,2 mm
    1 * 1 ST
    1 * 1 ST - 1 400,00 €
    1 400,00 €
    1 * 1 ST
       1 400,00 €
  • Socket S 51 with steel thread M 48×1,5, O.Ø 36×2,4 mm, I.Ø 31,2 mm
    1 * 1 ST
    1 * 1 ST - 1 730,00 €
    1 730,00 €
    1 * 1 ST
       1 730,00 €