Order Entry
Belgium
ContactUsLinkComponent
Capto™ adhere ImpRes multimodal chromatography media, Cytiva
Capto™ adhere ImpRes multimodal chromatography media, Cytiva
Supplier:  Cytiva
Certificates
undefined
Capto™ adhere ImpRes multimodal chromatography media, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • pH range:
    3 - 12
  • Matrix:
    High-flow agarose
  • Ligand:
    Multimodal strong anion exchanger
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    45 - 85 mg Mab/ml
  • Total ionic capacity:
    0,08 - 0,11 mmol Cl-/ml

Specifications

Frequently Bought Together