Safety containers for corrosives/acids, self-close
Self-close corrosive containers with stainless steel hardware are specifically designed to resist acids and caustics. Oval containers make the most of storage capacity and are marked in liters, quarts and gallons to permit accurate filling and dispensing.
HDPE, translucent.
- Spring-close cap provides a tight seal
- Flame arrester-free spout allows quick pouring of non-flammable corrosives
Frequently Bought Together