ContactUsLinkComponent
Hand crimper
Catalog # PERCN9302785
Supplier:  PerkinElmer
Hand crimper
Catalog # PERCN9302785
Supplier:  PerkinElmer
Specifications
  • Description:
    Hand crimper for chromatography vials, 20 mm
  • Pk:
    1
Specifications
About this item
Hand crimper for GC Headspace vials or LC large volume vials. It ensures sample integrity, particularly those containing volatile compounds.