Order Entry
Belgium
ContactUsLinkComponent
Sulphate standard solution, concentrate, 1000 mg SO₄, Titrisol®, Supelco®
Sulphate standard solution, concentrate, 1000 mg SO₄, Titrisol®, Supelco®
Catalog # 1.09872.0001
Supplier:  Merck
CAS Number:  
undefined
Sulphate standard solution, concentrate, 1000 mg SO₄, Titrisol®, Supelco®
Catalog # 1.09872.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Pk:
    1 Ampoul
  • CAS No.:
    7664-93-9
  • Description:
    Sulphate standard, Titrisol®
  • Seq:
    06
  • M-Clarity quality level:
    MQ100
  • Contents:
    H₂SO₄ in water

Specifications

About this item

Titrisol® standards are traceable to standard reference material from NIST.

Certifications: A batch-specific Certificate of Analysis is available.