Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
Sulphuric acid concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4700.
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Sulphuric acid concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4700.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    210 mL
  • Seq:
    2545

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589