Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid concentrate for dilution to 0.02 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sulphuric acid concentrate for dilution to 0.02 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4704.
Supplier:  Avantor
CAS Number:  
undefined
Sulphuric acid concentrate for dilution to 0.02 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4704.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589