Order Entry
Belgium
ContactUsLinkComponent
Sodium metabisulphite ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Irritant
Sodium metabisulphite ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3552-07
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Irritant
Sodium metabisulphite ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3552-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥ 97.0 %
  • Assay (Na₂S₂O₅):
    ≥97.0 %
  • Insoluble Matter:
    ≤0.005 %
  • Chloride (Cl):
    ≤0.05 %
  • Thiosulfate (S₂O₃):
    ≤0.05 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Iron (Fe):
    ≤0.002 %

Specification Test Results

About this item

 :
 :190.11 g/mol
Merck Index:13,08712
EINECS:231-673-0
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7681-57-4
Formula:Na₂S₂O₅
MDL Number:MFCD00167602