Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
Sodium hydroxide 2 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5633-02
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Sodium hydroxide 2 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5633-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    1.990 - 2.010
  • Protect from air to avoid absorption of carbon dioxide.:
    ≤5 ppm
  • Normality:
    1.990 - 2.010
  • Chloride (Cl):
    ≤ 5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤ 1 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.5 ppm

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548