Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide 0.5 N in methanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 0.5 N in methanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5631.1000
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 0.5 N in methanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5631.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.497 - 0.503
  • Normality:
    0.497 - 0.503
  • Chloride (Cl):
    ≤10 ppm
  • Nitrogen Compounds (as N):
    ≤1 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm

Specification Test Results

About this item

 :
 :56.11 g/mol
Density:0.80 g/cm³
Storage Temperature:Ambient
EINECS:215-181-3
UN:3286
ADR:3,II
 :GHS02
 :GHS06
 :GHS05
 :GHS08
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553