Human Isotype Control
Catalog # PIEA31154
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Control
- Antigen symbol:IgM
- Conjugation:Unconjugated
- Host:Human
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Seq:02
- Antibody fragment:whole
- Storage buffer:PBS, pH 7.6, with no preservative
- Storage temperature:+4 °C
- Concentration:11,0 mg/ml
- Tested applications:Blocking
- Purification:Ion-exchange chromatography.
- Pk:10 mg