Polyvinyl alcohol, M.W. ∼ 60000 for synthesis, fully hydrolysed, Sigma-Aldrich®
Catalog # 8.43866.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
Specification Test Results
- Melting Point:160 - 240 °C
- Bulk Density:400 - 600 kg/m3
- Density:0.4 - 0.6 g/cm3 (20 °C)
- Molecular Weight:60000
- Solubility:Insoluble at 20 °C
- Storage temperature:15...25 °C
- Degree of hydrolysis:≥98%
- dyn.Viscosity:9,0 - 11,0 mPa∙s
- Loss on drying (110 °C, 2 h):≤5,0%
- Ash:≤1,0%
- Ester value:15 - 25
Specification Test Results
About this item
Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®
:
:M-Clarity™ quality level = MQ200
CAS Number:9002-89-5
:25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922