Orders LinkContactUsLinkComponent
Spill control pallets, EcoPolyBlend™
Spill control pallets, EcoPolyBlend™
Spill control pallets, EcoPolyBlend™
Certificates
Spill control pallets, EcoPolyBlend™
Spill control pallets, EcoPolyBlend™
3 Options Available Below
Product Details & Documents
These pallets are made of up to 100% recycled polyethylene; dirt hiding black colour is 100% recycled, high visibility yellow is 35% recycled content. Fully compliant and quality engineered with a robust design, they withstand the toughest environments and are nearly impervious to chemical attack.
  • Independently validated by UL Environment
  • Seamless, robust design eliminates leaks
  • Excellent chemical compatibility and no corrosion
  • Sturdy and self-locating grates remove for easy cleaning
  • Square or space saving linear styles in multiple sizes offer storage versatility
Certifications: Meets or exceeds EPA, SPCC, NFPA 1, and the Int’l Fire Code.
Product Family Options
Product Information
  • Weight
    W×D×H
    Availability
    Price
  • 119-0376
    Spill control pallet, black, 250 liter sump capacity, for 2 drums, load capacity 1134 kg
    Spill control pallet, black, 250 liter sump capacity, for 2 drums, load capacity 1134 kg
    20,0 kg
    1245×635×457 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 369,00 €
    369,00 €
    1 * 1 ST
       369,00 €
  • 119-0378
    Spill control pallet, black, 276 liter sump capacity, for 4 square drums, load capacity 2268 kg
    Spill control pallet, black, 276 liter sump capacity, for 4 square drums, load capacity 2268 kg
    32,0 kg
    1245×1245×260 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 541,00 €
    541,00 €
    1 * 1 ST
       541,00 €
  • 119-0377
    Spill control pallet, black, 284 liter sump capacity, for 4 drums in line, load capacity 2268 kg
    Spill control pallet, black, 284 liter sump capacity, for 4 drums in line, load capacity 2268 kg
    35,0 kg
    2464×635×229 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 070,00 €
    1 070,00 €
    1 * 1 ST
       1 070,00 €
Accessories
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Ramp, for 4 drum square
    Ramp, for 4 drum square
    119-0391
    1245 x 838 x 267 mm
    Log in for availability
    1 * 1 ST - 492,00 €