Spécifications
- Antibody type:Primary
- Antigen name:Leucine-rich Repeat LGI family member 4
- Antigen symbol:LGI4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:163175
- Antigen synonyms:clp|Lgil3|Leucine-rich repeat LGI family member 4|MGC107385
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Spécifications
À propos de cet article
LGI4, also known as leucine-rich glioma-inactivated protein 4, is a 537 amino acid secreted glycosylated protein that is widely expressed, with highest levels found within the nervous system. Interestingly, siRNA knockdown studies of LGI4 expression in Schwann cells have been shown to result in the inhibition of myelination, thus suggesting that LGI4 is an essential component of myelin formation and axon segregation. LGI4 shares significant homology with its other family members, LGI1, LGI2 and LGI3. Significantly, mutations in the gene encoding LGI1 have been linked to human temporal lobe epilepsy and, given the sequence similarity of LGI4, it is likely that it also may be implicated in the pathology of seizures. LGI4 is localized subcellularly to the Golgi, ER and vesicles. There are two isoforms of LGI4 that are produced as a result of alternative splicing events.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: LGI4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat