Order Entry
Belgium
ContactUsLinkComponent
Anti-H2BFWT Rabbit Polyclonal Antibody
Anti-H2BFWT Rabbit Polyclonal Antibody
Catalog # BOSSBS-15401R
Supplier:  Bioss
Anti-H2BFWT Rabbit Polyclonal Antibody
Catalog # BOSSBS-15401R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    H2B histone family, member W, testis-specific
  • Antigen symbol:
    H2BFWT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    158983
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

H2BFWT (H2B histone family member W testis-specific) is a 175 amino acid nuclear membrane histone that belongs to the histone H2B family. In contrast to most H2B histones, H2BFWT does not contain the conserved C-terminal residue involved in monoubiquitination, but is structurally indistinguishable from conventional H2B histones. Also varying from other H2B histones, H2BFWT does not participate in the recruitment of chromosome condensation factors or in the assembly of mitotic chromsomes. Expressed in testis, H2BFWT is present in sperm cells and may be essential to telomere function.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: H2BFWT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human