Order Entry
Belgium
Orders LinkContactUsLinkComponent
Anti-MPP7 Rabbit Polyclonal Antibody

Anti-MPP7 Rabbit Polyclonal Antibody

100 μg
Catalog # BOSSBS-13737R
Supplier:   Bioss
Anti-MPP7 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13737R
Supplier:  Bioss
Anti-MPP7 Rabbit Polyclonal Antibody

100 μg

Price
668,00 €
Log in for availability
1 * 100 µl
Product Details & Documents

The MAGUK (membrane-associated guanylate kinase homologs) family of proteins contain multiple protein-binding domains and are involved in cell junction organization, tumor suppression, and signaling. The MAGUK family is divided into four subfamilies: DLG-like, ZO1-like, p55-like and LIN2-like. MPP7 (MAGUK p55 subfamily member 7), also known as palmitoylated membrane protein 7, is a 576 amino acid protein that belongs to the MAGUK family and assists in the assembly of protein complexes. A peripheral membrane protein that exists as a heterodimer, MPP7 enhances tight junction formation and epithelial cell polarity. MPP7 contains one SH3 domain, a guanylate kinase-like domain, two L27 domains and a single PDZ (DHR) domain. The gene encoding MPP7 maps to human chromosome 10p11.23

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MPP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Documents

Specifications
  • Size:
    100 μg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Purified by Protein A.
  • Antibody type:
    Primary
  • Form:
    liquid
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Cat. No.:
    BOSSBS-13737R
  • Reactivity:
    Human|Rat|Mouse
  • Antigen synonyms:
    Palmitoylated membrane protein 7.|FLJ32798|MAGUK p55 subfamily member 7|Membrane protein palmitoylated 7 MAGUK p55 subfamily member 7
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    MPP7
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.