Order Entry
Belgium
ContactUsLinkComponent
Anti-ATLF Rabbit Polyclonal Antibody
Anti-ATLF Rabbit Polyclonal Antibody
Catalog # BOSSBS-12564R
Supplier:  Bioss
Anti-ATLF Rabbit Polyclonal Antibody
Catalog # BOSSBS-12564R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Anthrax Toxin Lethal Factor
  • Antigen symbol:
    ATLF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    1158731
  • Antigen synonyms:
    Anthrax lethal factor|Anthrax lethal toxin endopeptidase component|LF|Anthrax LF|LEF_BACAN.|bacillus anthracis lethal factor|Lef
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

The protease enzyme Lethal Factor (LF) is one of the three proteins (LF, EF & PA) composing the anthrax toxin produced by Bacillus anthracis, a bacteria which can infect many mammalian species and that may be fatal. LF is not toxic by itself, but when associated with Protective Antigen (PA), can then gain entry to cells. Once inside the cell, LF then cleaves the N terminal of most dual specificity mitogen activated protein kinase kinases (MAPKKs or MAP2Ks) (except for MAP2K5). Cleavage invariably occurs within the N terminal proline rich region preceding the kinase domain, thus disrupting a sequence involved in directing specific protein protein interactions necessary for the assembly of signaling complexes. There may be other cytosolic targets of LF involved in cytotoxicity. The proteasome may mediate a toxic process initiated by LF in the cell cytosol involving degradation of unidentified molecules that are essential for macrophage homeostasis. This is an early step in LF intoxication, but it is downstream of the cleavage by LF of MEK1 or other putative substrates.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ATLF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others