Coaxial inserts for NMR precision sample tubes, SP Wilmad-LabGlass
Frequently Bought Together
VWR®, Beakers, Low Form
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
: DWK Life Sciences
epT.I.P.S.® Bulk - Pipette Tips
: EPPENDORF
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
: VWR Collection
SYLGARD® 184 Elastomer Kit
: DOW CORNING
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
NMR economy tubes, 5 mm, thin walled, SP Wilmad-LabGlass
: SP Industries
VWR®, Transfer Pipettes, High Performance
: VWR Collection