ContactUsLinkComponent
Schlenk sample tube, SP Wilmad-LabGlass
Schlenk sample tube, SP Wilmad-LabGlass
Catalog # WILMLG-10800-100
Supplier:  SP Industries
undefined
Schlenk sample tube, SP Wilmad-LabGlass
Catalog # WILMLG-10800-100
Supplier:  SP Industries
Specifications
  • Capacity:
    5 ml
  • Pk:
    1
Specifications
About this item
Tube incorporates a grease-free high vacuum 0 to 4 mm valve and is suited for pressure/vacuum applications. Ideal for reaction and storage of air-sensitive compounds. Sidearm can be connected to inert atmosphere/vacuum source or a manifold.