Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solution, colour coded, ready to use

Buffer solution, colour coded, ready to use

Supplier:   Bernd Kraft
Buffer solution, colour coded, ready to use
Supplier:  Bernd Kraft
Certificates
Buffer solution, colour coded, ready to use
Buffer solution, colour coded, ready to use

63 Options Available Below

Product Details & Documents
  • NIST traceable
  • Colour coded with choice of colours for pH values 4,00 and 7,00
Product Family Options

Product Information

  • pH value
    pH buffer type
    Colour
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • 2,0
    Citrate
    Yellow
    20 °C
    Plastic bottle
    0,5 L
    Log in for availability
    Log in for availability
    1 * 0,5 L
    1 * 0,5 L

  • 2,0
    Citrate
    Yellow
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 2,0
    Citrate
    Yellow
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 2,0
    Citrate
    Yellow
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 2,0
    Citrate
    Yellow
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 2,0
    Citrate
    Yellow
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 2,0
    Citrate
    Yellow
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 3,0
    Citrate
    Red
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • 3,0
    Citrate
    Red
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 3,0
    Citrate
    Red
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 3,0
    Citrate
    Red
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 3,0
    Citrate
    Red
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 3,0
    Citrate
    Red
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 3,0
    Citrate
    Red
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 4,0
    Citrate
    Blue
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • 4,0
    Citrate
    Blue
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 4,0
    Citrate
    Blue
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 4,0
    Citrate
    Blue
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 4,0
    Citrate
    Blue
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 4,0
    Citrate
    Blue
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 4,0
    Citrate
    Blue
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 4,0
    Citrate
    Red
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • 4,0
    Citrate
    Red
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 4,0
    Citrate
    Red
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 4,0
    Citrate
    Red
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 4,0
    Citrate
    Red
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 4,0
    Citrate
    Red
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 4,0
    Citrate
    Red
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 4,0
    Citrate
    Green
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • 4,0
    Citrate
    Green
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 4,0
    Citrate
    Green
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 4,0
    Citrate
    Green
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 4,0
    Citrate
    Green
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 4,0
    Citrate
    Green
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 4,0
    Citrate
    Green
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 7,0
    Phosphate
    Blue
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • 7,0
    Phosphate
    Blue
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 7,0
    Phosphate
    Blue
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 7,0
    Phosphate
    Blue
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 7,0
    Phosphate
    Blue
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 7,0
    Phosphate
    Blue
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 7,0
    Phosphate
    Blue
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 7,0
    Phosphate
    Green
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • 7,0
    Phosphate
    Green
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 7,0
    Phosphate
    Green
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 7,0
    Phosphate
    Green
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 7,0
    Phosphate
    Green
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 7,0
    Phosphate
    Green
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 7,0
    Phosphate
    Green
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 9,0
    Borate
    Blue
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • 9,0
    Borate
    Blue
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 9,0
    Borate
    Blue
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 9,0
    Borate
    Blue
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 9,0
    Borate
    Blue
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 9,0
    Borate
    Blue
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 9,0
    Borate
    Blue
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 10,0
    Borate
    Blue
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • 10,0
    Borate
    Blue
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 40,80 €
    40,80 €
    1 * 1 L
       40,80 €

  • 10,0
    Borate
    Blue
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,20 €
    45,20 €
    1 * 1 L
       45,20 €

  • 10,0
    Borate
    Blue
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 10,0
    Borate
    Blue
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 143,00 €
    143,00 €
    1 * 5 L
       143,00 €

  • 10,0
    Borate
    Blue
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 10,0
    Borate
    Blue
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €