Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

Buffer solutions, ready to use
Supplier:  Bernd Kraft
Certificates
Buffer solutions, ready to use
Buffer solutions, ready to use

105 Options Available Below

Product Details & Documents
  • NIST traceable

 

Product Family Options

Product Information

  • pH value
    pH buffer type
    Colour
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • 1,0
    HCl/KCl
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 1,0
    HCl/KCl
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 1,0
    HCl/KCl
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 1,0
    HCl/KCl
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 1,0
    HCl/KCl
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 1,0
    HCl/KCl
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 1,0
    HCl/KCl
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 13,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 2,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 2,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 2,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 2,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 2,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 2,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 2,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 3,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 3,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 3,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 3,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 3,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 3,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 3,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 4,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 4,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 4,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 4,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 4,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 4,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 4,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 4,66
    Acetate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 28,50 €
    28,50 €
    1 * 500 mL
       28,50 €

  • 4,66
    Acetate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 39,80 €
    39,80 €
    1 * 1 L
       39,80 €

  • 4,66
    Acetate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 44,20 €
    44,20 €
    1 * 1 L
       44,20 €

  • 4,66
    Acetate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 129,00 €
    129,00 €
    1 * 5 L
       129,00 €

  • 4,66
    Acetate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 4,66
    Acetate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 4,66
    Acetate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 5,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 5,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 5,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 5,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 5,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 5,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 5,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 5,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L

  • 5,50
    Acetate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 28,50 €
    28,50 €
    1 * 500 mL
       28,50 €

  • 5,50
    Acetate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 44,20 €
    44,20 €
    1 * 1 L
       44,20 €

  • 5,50
    Acetate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 129,00 €
    129,00 €
    1 * 5 L
       129,00 €

  • 5,50
    Acetate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 5,50
    Acetate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 189,00 €
    189,00 €
    1 * 10 L
       189,00 €

  • 5,50
    Acetate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 196,00 €
    196,00 €
    1 * 10 L
       196,00 €

  • 6,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 6,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 6,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 6,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 6,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 6,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 6,0
    Citrate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 7,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 7,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 35,50 €
    35,50 €
    1 * 1 L
       35,50 €

  • 7,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 7,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 7,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 7,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 7,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 8,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 8,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 8,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 8,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 8,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 8,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 8,0
    Phosphate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 9,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 9,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 9,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 9,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 9,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 9,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 9,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 10,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 10,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 10,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 10,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 10,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 10,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 10,0
    Borate
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 11,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 11,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 11,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 11,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 11,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 11,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 11,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 12,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 12,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 12,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 12,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 12,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 12,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €

  • 12,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • 13,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 24,70 €
    24,70 €
    1 * 500 mL
       24,70 €

  • 13,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,80 €
    36,80 €
    1 * 1 L
       36,80 €

  • 13,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,30 €
    41,30 €
    1 * 1 L
       41,30 €

  • 13,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • 13,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 139,00 €
    139,00 €
    1 * 5 L
       139,00 €

  • 13,0
    Glycine
    Colourless
    20 °C
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 182,00 €
    182,00 €
    1 * 10 L
       182,00 €